3-(Methylthio)propionyl chloride 95%

Lieferant: Apollo Scientific

Synonyme: 3-(Methylmercapto)propionyl Chloride

OR921893-5G
APOSOR921893-5GEA 78.3 EUR
APOSOR921893-5G
3-(Methylthio)propionyl chloride 95%
3-(Methylthio)propionyl chloride
Formel: C₄H₇ClOS
Molecular Weight: 138.61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059482
CAS-Nummer: 7031-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR