4-Brom-2-(2-methyl-2-propanyl)phenol

Lieferant: Apollo Scientific
Achtung

OR921949-5G
APOSOR921949-5GEA 68.3 EUR
APOSOR921949-5G
4-Brom-2-(2-methyl-2-propanyl)phenol
4-Brom-2-(2-methyl-2-propanyl)phenol
Formel: C₁₀H₁₃BrO
Lagertemperatur: Kühlschrank
MDL: MFCD00192645
CAS-Nummer: 10323-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR