4-Bromo-5-chloro-2,3-dihydropyridazin-3-one

Lieferant: Apollo Scientific

OR921977-1G
APOSOR921977-1GEA 923 EUR
APOSOR921977-1G
4-Bromo-5-chloro-2,3-dihydropyridazin-3-one
4-Bromo-5-chloro-2,3-dihydropyridazin-3-one
Formel: C₄H₂BrClN₂O
Molecular Weight: 209,428 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106328
CAS-Nummer: 119729-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR