3-Methoxycyclobutane-1-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-Methoxycyclobutanecarboxylic acid

OR922058-1G
APOSOR922058-1GEA 423 EUR
APOSOR922058-1G
3-Methoxycyclobutane-1-carboxylic acid
3-Methoxycyclobutane-1-carboxylic acid
Formel: C₆H₁₀O₃
Molecular Weight: 130,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705860
CAS-Nummer: 480450-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR