7-Aminothieno[2,3-b]pyrazine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Thieno[2,3-b]pyrazin-7-amine
OR9221-250MG
APOSOR9221-250MGEA
105
EUR
APOSOR9221-250MG
7-Aminothieno[2,3-b]pyrazine 95%
7-Aminothieno[2,3-b]pyrazine
Formel:
C₆H₅N₃S Molecular Weight: 151,189 g/mol Schmelzpunkt: 87,5…89.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271970 CAS-Nummer: 59944-75-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...