2-(Hexahydro-1H-pyrrolizin-7a-yl)ethanamine 95%

Lieferant: Apollo Scientific

OR922115-1G
APOSOR922115-1GEA 574 EUR
APOSOR922115-1G
2-(Hexahydro-1H-pyrrolizin-7a-yl)ethanamine 95%
2-(Hexahydro-1H-pyrrolizin-7a-yl)ethanamine
Formel: C₉H₁₈N₂
Molecular Weight: 154,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702871
CAS-Nummer: 78449-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR