Methyl 3-(2-pyridyl)propiolate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-(2-Pyridyl)propiolate
OR922119-1G
APOSOR922119-1GEA
246
EUR
APOSOR922119-1G
Methyl 3-(2-pyridyl)propiolate 95%
Methyl 3-(2-pyridyl)propiolate
Formel:
C₉H₇NO₂ Molecular Weight: 161,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02646628 CAS-Nummer: 72764-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...