(2R)-2-(Methoxymethyl)azetidine

Lieferant: Apollo Scientific

Synonyme: (R)-2-(Methoxymethyl)azetidine

OR922129-1G
APOSOR922129-1GEA 687 EUR
APOSOR922129-1G
(2R)-2-(Methoxymethyl)azetidine
(2R)-2-(Methoxymethyl)azetidine
Formel: C₅H₁₁NO
Molecular Weight: 101.147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977227
CAS-Nummer: 935668-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR