Methyl cis-4-hydroxycyclohexanecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: cis-Methyl 4-hydroxycyclohexanecarboxylate

OR922132-1G
APOSOR922132-1GEA 30.6 EUR
APOSOR922132-1G
Methyl cis-4-hydroxycyclohexanecarboxylate 95%
Methyl cis-4-hydroxycyclohexanecarboxylate
Formel: C₈H₁₄O₃
Molecular Weight: 158,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441962
CAS-Nummer: 3618-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR