Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%

Lieferant: Apollo Scientific

OR922142-1G
APOSOR922142-1GEA 82.8 EUR
APOSOR922142-1G
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate 95%
Ethyl 1H-pyrrolo[2,3-c]pyridine-2-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11506124
CAS-Nummer: 24334-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR