1-(8-Hydroxyquinolin-5-yl)ethanone 95%

Lieferant: Apollo Scientific

OR922155-1G
APOSOR922155-1GEA 406 EUR
APOSOR922155-1G
1-(8-Hydroxyquinolin-5-yl)ethanone 95%
1-(8-Hydroxyquinolin-5-yl)ethanone
Formel: C₁₁H₉NO₂
Molecular Weight: 187.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086061
CAS-Nummer: 2598-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR