Di(2-pyridyl)ketonoxim 95%
Lieferant: Apollo Scientific
Synonyme:
2,2'-Dipyridyl ketoxime
OR922157-1G
OR922157-5G
APOSOR922157-1GEA
219
EUR
APOSOR922157-1G
APOSOR922157-5G
Di(2-pyridyl)ketonoxim 95%
Di(2-pyridyl)ketonoxim
Formel:
C₁₁H₉N₃O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006311 CAS-Nummer: 1562-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...