3-Nitro-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 3-Nitro-7-azaindole

OR922183-5G
APOSOR922183-5GEA 145 EUR
APOSOR922183-5G
3-Nitro-1H-pyrrolo[2,3-b]pyridine 95%
3-Nitro-1H-pyrrolo[2,3-b]pyridine
Formel: C₇H₅N₃O₂
Molecular Weight: 163.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08272225
CAS-Nummer: 23709-47-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR