1-Nitro-2-naphthol 95%

Lieferant: Apollo Scientific

OR922194-1G OR922194-250MG
APOSOR922194-1GEA 0 EUR
APOSOR922194-1G APOSOR922194-250MG
1-Nitro-2-naphthol 95%
1-Nitro-2-naphthol
Formel: C₁₀H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00021461
CAS-Nummer: 550-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR