5-Methyl-2-furonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 5-Methylfuran-2-carbonitrile

OR922196-1G
APOSOR922196-1GEA 77 EUR
APOSOR922196-1G
5-Methyl-2-furonitrile 95%
5-Methyl-2-furonitrile
Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093782
CAS-Nummer: 13714-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR