1-Methyl-1H-indole-4-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-4-indolecarboxylic Acid

OR9222-1G OR9222-5G OR9222-250MG
APOSOR9222-1GEA 58 EUR
APOSOR9222-1G APOSOR9222-5G APOSOR9222-250MG
1-Methyl-1H-indole-4-carboxylic acid 95%
1-Methyl-1H-indole-4-carboxylic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175.19 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08435874
CAS-Nummer: 90924-06-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR