4-Amino-2-pyridincarbonitril 98%

Lieferant: Apollo Scientific

Synonyme: 4-Aminopicolinonitrile

OR922209-1G OR922209-250MG
APOSOR922209-1GEA 128 EUR
APOSOR922209-1G APOSOR922209-250MG
4-Amino-2-pyridincarbonitril 98%
4-Amino-2-pyridincarbonitril
Formel: C₆H₅N₃
Molecular Weight: 119.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374351
CAS-Nummer: 98139-15-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR