Methyl imidazo[1,5-a]pyridine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR922221-1G
APOSOR922221-1GEA 647 EUR
APOSOR922221-1G
Methyl imidazo[1,5-a]pyridine-1-carboxylate 95%
Methyl imidazo[1,5-a]pyridine-1-carboxylate
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15144579
CAS-Nummer: 1039356-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR