4-Chloro-3-methoxy-2-methylpyridine 95%

Lieferant: Apollo Scientific

OR922245-1G
APOSOR922245-1GEA 92.8 EUR
APOSOR922245-1G
4-Chloro-3-methoxy-2-methylpyridine 95%
4-Chloro-3-methoxy-2-methylpyridine
Formel: C₇H₈ClNO
Molecular Weight: 157.6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16610605
CAS-Nummer: 107512-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR