1,4-Dioxaspiro[4.5]decan-7-one 95%

Lieferant: Apollo Scientific

OR922251-1G
APOSOR922251-1GEA 203 EUR
APOSOR922251-1G
1,4-Dioxaspiro[4.5]decan-7-one 95%
1,4-Dioxaspiro[4.5]decan-7-one
Formel: C₈H₁₂O₃
Molecular Weight: 156,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16039576
CAS-Nummer: 4969-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR