tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate 95%

Lieferant: Apollo Scientific

OR922315-1G
APOSOR922315-1GEA 487 EUR
APOSOR922315-1G
tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate 95%
tert-Butyl 5-methyl-3,6-dihydro-2H-pyridine-1-carboxylate
Formel: C₁₁H₁₉NO₂
Molecular Weight: 197,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19703948
CAS-Nummer: 1211531-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR