Phenyltrimethoxysilan 98%

Lieferant: Apollo Scientific

OR922360-100G OR922360-500G
APOSOR922360-100GEA 0 EUR
APOSOR922360-100G APOSOR922360-500G
Phenyltrimethoxysilan 98%
Phenyltrimethoxysilan
Formel: C₉H₁₄O₃Si
Lagertemperatur: Raumtemperatur
MDL: MFCD00025689
CAS-Nummer: 2996-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR