4-Methoxy-1H-indazole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR922400-1G
APOSOR922400-1GEA 422 EUR
APOSOR922400-1G
4-Methoxy-1H-indazole-3-carboxylic acid
4-Methoxy-1H-indazole-3-carboxylic acid
Formel: C₉H₈N₂O₃
Molecular Weight: 192,171 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07371558
CAS-Nummer: 865887-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR