Methyl 2-cyclohexyl-2-oxoacetate 95%

Lieferant: Apollo Scientific

OR922435-1G
APOSOR922435-1GEA 46.4 EUR
APOSOR922435-1G
Methyl 2-cyclohexyl-2-oxoacetate 95%
Methyl 2-cyclohexyl-2-oxoacetate
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24688272
CAS-Nummer: 62783-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR