2-Hydroxy-N-methylbenzamide 95%

Lieferant: Apollo Scientific

Synonyme: N-Methylsalicylamide

OR922451-5G
APOSOR922451-5GEA 177 EUR
APOSOR922451-5G
2-Hydroxy-N-methylbenzamide 95%
2-Hydroxy-N-methylbenzamide
Formel: C₈H₉NO₂
Molecular Weight: 151.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045757
CAS-Nummer: 1862-88-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR