2-Cyclobutylethan-1-amine

Lieferant: Apollo Scientific

OR922462-1G
APOSOR922462-1GEA 811 EUR
APOSOR922462-1G
2-Cyclobutylethan-1-amine
2-Cyclobutylethan-1-amine
Formel: C₆H₁₃N
Molecular Weight: 99,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09924416
CAS-Nummer: 60637-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR