Sodium hydroxymethanesulphinate 95%

Lieferant: Apollo Scientific
Achtung

OR922468-100G OR922468-25G
APOSOR922468-100GEA 75.9 EUR
APOSOR922468-100G APOSOR922468-25G
Sodium hydroxymethanesulphinate 95%
Sodium hydroxymethanesulphinate
Formel: CH₃NaO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00040426
CAS-Nummer: 149-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR