2-(Hydroxymethyl)cyclobutan-1-one

Lieferant: Apollo Scientific

Synonyme: 2-(Hydroxymethyl)cyclobutanone

OR922566-1G
APOSOR922566-1GEA 683 EUR
APOSOR922566-1G
2-(Hydroxymethyl)cyclobutan-1-one
2-(Hydroxymethyl)cyclobutan-1-one
Formel: C₅H₈O₂
Molecular Weight: 100.116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106469
CAS-Nummer: 23107-52-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR