3,4,5-Trichloropyridin-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3,4,5-trichloropyridine

OR922583-1G
APOSOR922583-1GEA 72.5 EUR
APOSOR922583-1G
3,4,5-Trichloropyridin-2-amine 95%
3,4,5-Trichloropyridin-2-amine
Formel: C₅H₃Cl₃N₂
Molecular Weight: 197.45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705655
CAS-Nummer: 55933-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR