2,3-Dimethyl-1H-indole-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR9226-1G
APOSOR9226-1GEA 292 EUR
APOSOR9226-1G
2,3-Dimethyl-1H-indole-5-carboxylic acid 97%
2,3-Dimethyl-1H-indole-5-carboxylic acid
Formel: C₁₁H₁₁NO₂
Schmelzpunkt: 239…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00464053
CAS-Nummer: 14844-73-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR