7-Chlorimidazo[4,5-c]pyridin 95%

Lieferant: Apollo Scientific
Achtung

OR922610-250MG
APOSOR922610-250MGEA 116 EUR
APOSOR922610-250MG
7-Chlorimidazo[4,5-c]pyridin 95%
7-Chlorimidazo[4,5-c]pyridin
Formel: C₆H₄ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11501262
CAS-Nummer: 5975-13-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR