N-(4-Methoxyphenyl)formamid 95%

Lieferant: Apollo Scientific

Synonyme: N-(4-Methoxyphenyl)formamide, N-Formyl-4-methoxyaniline

OR922686-5G
APOSOR922686-5GEA 115 EUR
APOSOR922686-5G
N-(4-Methoxyphenyl)formamid 95%
N-(4-Methoxyphenyl)formamid
Formel: C₈H₉NO₂
Molecular Weight: 151.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00086774
CAS-Nummer: 5470-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR