5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 95%
Lieferant: Apollo Scientific
OR922722-1G
APOSOR922722-1GEA
191
EUR
APOSOR922722-1G
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 95%
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine
Formel:
C₇H₆N₄O Lagertemperatur: Raumtemperatur |
MDL:
MFCD01710636 CAS-Nummer: 5711-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...