Natrium-L(+)-ascorbat 98%

Lieferant: Apollo Scientific

OR922808-500G
APOSOR922808-500GEA 0 EUR
APOSOR922808-500G
Natrium-L(+)-ascorbat 98%
Natrium-L(+)-ascorbat
Formel: C₆H₇NaO₆
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082340
CAS-Nummer: 134-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR