(1S)-(+)-Campher-10-sulfonsäure 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
D(+)-Campher-10-sulfonsäure, (1S)-(+)-2-Oxobornan-10-sulfonsäure
OR922840-100G
APOSOR922840-100GEA
56.7
EUR
APOSOR922840-100G
(1S)-(+)-Campher-10-sulfonsäure 98%
(1S)-(+)-Campher-10-sulfonsäure
Formel:
C₁₀H₁₆O₄S Molecular Weight: 232,3 g/mol Schmelzpunkt: 196…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064157 CAS-Nummer: 3144-16-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...