(R)-(+)-3-Cyclohexenecarboxylic acid 98%

Lieferant: Apollo Scientific

OR922857-100G
APOSOR922857-100GEA 88.6 EUR
APOSOR922857-100G
(R)-(+)-3-Cyclohexenecarboxylic acid 98%
(R)-(+)-3-Cyclohexenecarboxylic acid
Formel: C₇H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08275445
CAS-Nummer: 5709-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR