N-(Cyclopropylmethyl)cyclopropanamine 95%

Lieferant: Apollo Scientific

OR922970-1G
APOSOR922970-1GEA 719 EUR
APOSOR922970-1G
N-(Cyclopropylmethyl)cyclopropanamine 95%
N-(Cyclopropylmethyl)cyclopropanamine
Formel: C₇H₁₃N
Molecular Weight: 111,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11212563
CAS-Nummer: 215522-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR