2-(Dimethylamino)ethyl-acetat 95%

Lieferant: Apollo Scientific

Synonyme: Acetic Acid 2-(Dimethylamino)ethyl Ester

OR922974-100G OR922974-25G
APOSOR922974-100GEA 435 EUR
APOSOR922974-100G APOSOR922974-25G
2-(Dimethylamino)ethyl-acetat 95%
2-(Dimethylamino)ethyl acetate
Formel: C₆H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00041912
CAS-Nummer: 1421-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR