(4-Methylpyridin-3-yl)methanol 95%
Lieferant: Apollo Scientific
OR922994-1G
APOSOR922994-1GEA
148
EUR
APOSOR922994-1G
(4-Methylpyridin-3-yl)methanol 95%
(4-Methylpyridin-3-yl)methanol
Formel:
C₇H₉NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD01444898 CAS-Nummer: 4664-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...