Methyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-7-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR922998-1G
APOSOR922998-1GEA 647 EUR
APOSOR922998-1G
Methyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-7-carboxylate 95%
Methyl 5,6,7,8-tetrahydroimidazo[1,2-a]pyridine-7-carboxylate
Formel: C₉H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22373387
CAS-Nummer: 1623060-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR