3-Benzyl-3,8-diazabicyclo[3.2.1]octane
Lieferant: Apollo Scientific
OR923010-1G
APOSOR923010-1GEA
221
EUR
APOSOR923010-1G
3-Benzyl-3,8-diazabicyclo[3.2.1]octane
3-Benzyl-3,8-diazabicyclo[3.2.1]octane
Formel:
C₁₃H₁₈N₂ Molecular Weight: 202,295 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20441694 CAS-Nummer: 67571-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...