1-Piperidinecarbonyl chloride 96%

Lieferant: Apollo Scientific

OR923036-25G
APOSOR923036-25GEA 0 EUR
APOSOR923036-25G
1-Piperidinecarbonyl chloride 96%
1-Piperidinecarbonyl chloride
Formel: C₆H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00973554
CAS-Nummer: 13939-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR