N-(2-Hydroxyethyl)phthalimid 98%
Lieferant: Apollo Scientific
Synonyme:
N-(2-Hydroxyethyl)phthalsäureimid, N-(2-Hydroxyethyl)-1,2-benzoldicarbonsäureimid
OR923052-100G
OR923052-1KG
OR923052-500G
APOSOR923052-100GEA
53.8
EUR
APOSOR923052-100G
APOSOR923052-1KG
APOSOR923052-500G
N-(2-Hydroxyethyl)phthalimid 98%
N-(2-Hydroxyethyl)phthalimid
Formel:
C₁₀H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005903 CAS-Nummer: 3891-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...