N-(2-Hydroxyethyl)phthalimid 98%

Lieferant: Apollo Scientific

Synonyme: N-(2-Hydroxyethyl)phthalsäureimid, N-(2-Hydroxyethyl)-1,2-benzoldicarbonsäureimid

OR923052-100G OR923052-1KG OR923052-500G
APOSOR923052-100GEA 53.8 EUR
APOSOR923052-100G APOSOR923052-1KG APOSOR923052-500G
N-(2-Hydroxyethyl)phthalimid 98%
N-(2-Hydroxyethyl)phthalimid
Formel: C₁₀H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00005903
CAS-Nummer: 3891-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR