4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%

Lieferant: Apollo Scientific

OR923083-1G OR923083-250MG
APOSOR923083-1GEA 0 EUR
APOSOR923083-1G APOSOR923083-250MG
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride 95%
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride
Formel: C₆H₁₂ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD22123337
CAS-Nummer: 95019-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR