4-Benzyloxy-3,3-dimethylbut-1-yne 95%

Lieferant: Apollo Scientific

OR923088-1G
APOSOR923088-1GEA 487 EUR
APOSOR923088-1G
4-Benzyloxy-3,3-dimethylbut-1-yne 95%
4-Benzyloxy-3,3-dimethylbut-1-yne
Formel: C₁₃H₁₆O
Molecular Weight: 188,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23106327
CAS-Nummer: 1092536-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR