2-Butylanilin 95%
Lieferant: Apollo Scientific
OR923102-5G
APOSOR923102-5GEA
359
EUR
APOSOR923102-5G
2-Butylanilin 95%
2-Butylanilin
Formel:
C₁₀H₁₅N Molecular Weight: 149,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00130033 CAS-Nummer: 2696-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...