(±)-1-Amino-2-propanol 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (±)-Isopropanolamin, (±)-1-Aminopropan-2-ol

OR923138-100G
APOSOR923138-100GEA 21.9 EUR
APOSOR923138-100G
(±)-1-Amino-2-propanol 95%
(±)-1-Amino-2-propanol
Formel: C₃H₉NO
Molecular Weight: 75,11 g/mol
Flammpunkt: 105 °C
MDL: MFCD00008139
CAS-Nummer: 78-96-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR