1-Tosylazetidin-3-ol

Lieferant: Apollo Scientific
Achtung

OR923177-250MG
APOSOR923177-250MGEA 405 EUR
APOSOR923177-250MG
1-Tosylazetidin-3-ol
1-Tosylazetidin-3-ol
Formel: C₁₀H₁₃NO₃S
Schmelzpunkt: 103…107 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00963433
CAS-Nummer: 154010-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR