9-Azabicyclo[3.3.1]nonan-3-ol 95%

Lieferant: Apollo Scientific

OR923178-1G
APOSOR923178-1GEA 400 EUR
APOSOR923178-1G
9-Azabicyclo[3.3.1]nonan-3-ol 95%
9-Azabicyclo[3.3.1]nonan-3-ol
Formel: C₈H₁₅NO
Molecular Weight: 141.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22631510
CAS-Nummer: 209733-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR